Learn More
inositol polyphosphate-5-phosphatase, 40kDa, Mouse, Clone: 3D8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant INPP5A.
Supplier: Abnova Corporation H00003632M05
Description
The protein encoded by this gene is a membrane-associated type I inositol 1,4,5-trisphosphate (InsP3) 5-phosphatase. InsP3 5-phosphatases hydrolyze Ins(1,4,5)P3, which mobilizes intracellular calcium and acts as a second messenger mediating cell responses to various stimulation. [provided by RefSeq
Sequence: YFNQEVFRDNNGTALLEFDKELSVFKDRLYELDISFPPSYPYSEDARQGEQYMNTRCPAWCDRILMSPSAKELVLRVSVCCPSPGHRGMWSAGSGLAQPWSpecifications
inositol polyphosphate-5-phosphatase, 40kDa | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_005539 | |
INPP5A | |
INPP5A (NP_005530, 288 a.a. ∼ 387 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody |
ELISA, Immunoprecipitation, Western Blot | |
3D8 | |
Mouse monoclonal antibody raised against a partial recombinant INPP5A. | |
INPP5A | |
5PTASE/DKFZp434A1721/MGC116947/MGC116949 | |
Mouse | |
Affinity Purified | |
RUO | |
3632 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.