Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INS, Mouse, Clone: 7C8, Abnova™

Catalog No. 89123198 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-198 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-198 Supplier Abnova Corporation Supplier No. H00003630M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a KLH-conjugated peptide spanning 48 - 62 amino acid sequence of human Proinsulin.

After removal of the precursor signal peptide, proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds. Binding of this mature form of insulin to the insulin receptor (INSR) stimulates glucose uptake. A variety of mutant alleles with changes in the coding region have been identified. There is a read-through gene, INS-IGF2, which overlaps with this gene at the 5' region and with the IGF2 gene at the 3' region. [provided by RefSeq]

Sequence: CFFYTPKTRREAEDLQ

Specifications

Antigen INS
Applications ELISA
Classification Monoclonal
Clone 7C8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a KLH-conjugated peptide spanning 48 ∼ 62 amino acid sequence of human Proinsulin.
Formulation PBS with no preservative; pH 7.4
Gene INS
Gene Accession No. NM_000207
Gene Alias ILPR/IRDN
Gene Symbols INS
Host Species Mouse
Immunogen KLH-conjugated human Proinsulin peptide (NP_000198, 48 a.a. ∼ 62 a.a)
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3630
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.