Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

insulin receptor-related receptor, Mouse, Clone: 5C6, Abnova™

Catalog No. 89000756 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-756 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-756 Supplier Abnova Corporation Supplier No. H00003645M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant INSRR.

Sequence: LYLNDYCHRGLRLPTSNNDPRFDGEDGDPEAEMESDCCPCQHPPPGQVLPPLEAQEASFQKKFENFLHNAITIPISPWKVTSINKSPQRDSGRHRRAAGPLRLGGNSSDF

Specifications

Antigen insulin receptor-related receptor
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5C6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant INSRR.
Formulation PBS with no preservative; pH 7.4
Gene INSRR
Gene Accession No. NM_014215
Gene Alias IRR
Gene Symbols INSRR
Host Species Mouse
Immunogen INSRR (NP_055030, 651 a.a. ∼ 760 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Kinases
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 3645
Target Species Human, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.