Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin Mouse anti-Human, Clone: MAB1, MilliporeSigma™

Catalog No. 051108MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-110-8MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-110-8MI Supplier MilliporeSigma Supplier No. 051108
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Insulin Antibody, clone1 is an antibody against Insulin for use in immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Insulin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MAB1
Conjugate Unconjugated
Formulation The monoclonal is presented as 1mg in 1ml phosphate buffered saline containing 10mM sodium azide. We recommend that each laboratory determine an optimum working titre for use in its particular application.
Gene Accession No. P01308
Gene Symbols ILPR, IRDN, OTTHUMP00000011161 , OTTHUMP00000011162, OTTHUMP00000196038, insulin, proinsulin
Host Species Mouse
Immunogen Biosynthetic human insulin
Purification Method Affinity Purified
Quantity 1 mg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.