Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin-like Growth Factor-1 Receptor Mouse anti-Human, Clone: 24-31, MilliporeSigma™

Catalog No. MAB1120MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1120MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1120MI Supplier MilliporeSigma Supplier No. MAB1120
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Insulin-like Growth Factor-1 Receptor Antibody, α-subunit, clone 24-31 detects level of Insulin-like Growth Factor-1 Receptor and has been published and validated for use in immunohistochemistry (paraffin) and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Insulin-like Growth Factor-1 Receptor
Applications Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone 24-31
Conjugate Unconjugated
Formulation Purified from ascites fluid by Protein G chromatography. Liquid in 10mM PBS, pH 7.4, with 0.2% BSA and 0.09% sodium azide.
Gene Accession No. NM_000875.3
Gene Symbols IGF1R, MGC142170, MGC18216, IGFIR, JTK13, MGC142172, CD221, EC 2.7.10.1 [Contains: Insulin-like growth factor 1 receptor alpha chain, Insulin-like growth factor 1 receptor beta chain].
Host Species Mouse
Immunogen IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.