Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin Receptor Mouse anti-Bovine, Human, Pig, Sheep, Clone: 47-9, MilliporeSigma™

Catalog No. MAB1137MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1137MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1137MI Supplier MilliporeSigma Supplier No. MAB1137

Mouse Monoclonal Antibody

Anti-Insulin Receptor Antibody, α subunit, clone 47-9 detects level of Insulin Receptor and has been published and validated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Insulin Receptor
Applications Functional Assay
Classification Monoclonal
Clone 47-9
Conjugate Unconjugated
Formulation Liquid in 10mM PBS, pH 7.4, with 0.2% BSA and 0.09% sodium azide.
Gene Accession No. NM_000208.2; NM_001079817.1
Gene Symbols INSR, CD220, HHF5, IR, EC 2.7.10.1 [Contains: Insulin receptor subunit alpha, Insulin receptor subunit beta].
Host Species Mouse
Immunogen IM-9 lymphocytes followed by purified insulin receptor.
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Human, Pig, Sheep
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.