Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IDE Monoclonal Antibody (334501)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524064
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene may belong to a protease family responsible for intercellular peptide signalling. Though its role in the cellular processing of insulin has not yet been defined, insulin-degrading enzyme is thought to be involved in the termination of the insulin response.
Specifications
| IDE | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P14735 | |
| IDE | |
| Sf 21-derived recombinant human Insulysin/IDE Met42-Leu1019. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| 334501 | |
| Unconjugated | |
| IDE | |
| 1300012G03Rik; 4833415K22Rik; AA675336; Abeta-degrading protease; AI507533; FLJ35968; Ide; INSDEGM; Insulin degrading enzyme; insulin protease; insulinase; Insulin-degrading enzyme; Insulysin; zgc:162603 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3416 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction