Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin Β1 (CD29) conjugate Rat anti-Mouse, FITC, Clone: MB1.2, MilliporeSigma™

Catalog No. FCMAB269FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB269FMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB269FMI Supplier MilliporeSigma Supplier No. FCMAB269F
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This Milli-Mark™ Anti-Integrin β1 (CD29)-FITC conjugate Antibody, clone MB1.2 is validated for use in FC for the detection of Integrin β1 (CD29) conjugate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin β1 (CD29)
Applications Flow Cytometry
Classification Monoclonal
Clone MB1.2
Conjugate FITC
Formulation Purified rat monoclonal IgG2a conjugated to FITC in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Accession No. NP_002202
Gene Symbols ITGB1, GPIIA, OTTHUMP00000063731, OTTHUMP00000046253, OTTHUMP00000063733, VLAB, OTTHUMP00000063732, OTTHUMP00000046253, MDF2, FNRB, CD29, MSK12, VLA-BETA
Host Species Rat
Purification Method Protein A Purified
Quantity 100 Tests
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for up to 6 months. Protect from light.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.