Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin Β1 Mouse anti-Human, Clone: 12G10, MilliporeSigma™

Catalog No. MAB2247IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2247IMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2247IMI Supplier MilliporeSigma Supplier No. MAB2247I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Integrin β1 Antibody, clone 12G10 is validated for use in Western Blotting, Flow Cytometry, Immunofluorescence for the detection of Integrin β1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin β1
Applications Flow Cytometry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 12G10
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in PBS containing 0.1% sodium azide.
Gene Accession No. NP_002283
Gene Symbols ITGB1, FNRB, MDF2, MSK12
Host Species Mouse
Immunogen Purified human β1 Integrin preparation from HT1080 fibrosarcoma cell extract
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.