Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin Β1 Mouse anti-Human, Clone: B44, MilliporeSigma™

Catalog No. MAB2259ZMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2259ZMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2259ZMI Supplier MilliporeSigma Supplier No. MAB2259Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Integrin β1 Antibody, activated, clone B44, azide free is validated for use in flow cytometry, immunohistochemistry, immunoprecipitation, FUNC, CULT, western blotting for the detection of Integrin β1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin β1
Applications Flow Cytometry, Functional Assay, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone B44
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid PBS, containing no preservatives.
Gene Accession No. NM_002292.3
Gene Symbols LAMB2, S-laminin, LAMS
Host Species Mouse
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.