Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin α1 Mouse anti-Human, Clone: FB12, MilliporeSigma™

Catalog No. MAB1973MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1973MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1973MI Supplier MilliporeSigma Supplier No. MAB1973
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Integrin α1 Antibody, clone FB12 is validated for use in ELISA, flow cytometry, immunocytochemistry, immunohistochemistry, immunoprecipitation, FUNC, western blotting for the detection of Integrin α1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin α1
Applications ELISA, Flow Cytometry, Functional Assay, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone FB12
Conjugate Unconjugated
Formulation Purified immunoglobulin in 0.02M PB, pH 7.6, 0.25M NaCl containing 0.1% sodium azide.
Gene Accession No. NM_181501.1
Gene Symbols ITGA1, CD49a, VLA1, VLA-1
Host Species Mouse
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.