Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin Β1 Mouse anti-Human, Pig, Clone: HB1.1, MilliporeSigma™

Catalog No. MAB2000MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2000MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2000MI Supplier MilliporeSigma Supplier No. MAB2000
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Integrin β1 Antibody, clone HB1.1 detects level of Integrin β1 and has been published and validated for use in flow cytometry, immunoprecipitation, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin β1
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone HB1.1
Conjugate Unconjugated
Formulation Purified immunoglobulin from Protein A chromatography. Liquid in 0.02M PB pH 7.6, 0.25M NaCl with 0.1% sodium azide.
Gene Accession No. NM_002211.2; NM_033666.1; NM_033667.1; NM_033668.1; NM_033669.1; NM_133376.1
Gene Symbols ITGB1, GPIIA, OTTHUMP00000063731, OTTHUMP00000046253, OTTHUMP00000063733, VLAB, OTTHUMP00000063732, MDF2, FNRB, CD29, MSK12
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Pig
Content And Storage 2-8°C for up to 6 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.