Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin Β1 Mouse anti-Human, Clone: JB1A (a.k.a. J10), MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1965
Description
This Anti-Integrin β1 Antibody, a.a. 82-87, clone JB1A (a.k.a. J10) is validated for use in flow cytometry, immunohistochemistry, immunoprecipitation, FUNC, western blotting for the detection of Integrin β1.
Specifications
Integrin β1 | |
Monoclonal | |
Unconjugated | |
NM_002211.2; NM_033666.1; NM_033667.1; NM_033668.1; NM_033669.1; NM_133376.1 | |
Mouse | |
Unpurified | |
Primary | |
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Flow Cytometry, Functional Assay, Immunohistochemistry, Immunoprecipitation, Western Blot | |
JB1A (a.k.a. J10) | |
Ascites fluid containing no preservatives. | |
ITGB1, GPIIA, OTTHUMP00000063731, OTTHUMP00000046253, OTTHUMP00000063733, VLAB, OTTHUMP00000063732, MDF2, FNRB, CD29, MSK12 | |
Purified beta 1 integrin and Jurkat cells | |
100 μL | |
Human | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction