Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin Β1 Rat anti-Mouse, Rat, Human, Clone: MB1.2, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MAB1997
Description
Anti-Integrin β1 Antibody, clone MB1.2 is an antibody against Integrin β1 for use in flow cytometry, immunohistochemistry, immunoprecipitation and western blotting.
Specifications
Integrin β1 | |
Monoclonal | |
Unconjugated | |
NP_002202 | |
Rat | |
100 μg | |
Mouse, Rat | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
MB1.2 | |
Purified rat monoclonal IgG2κ liquid in buffer containing 0.02M phosphate buffer, pH 7.6, 0.25M NaCl with 0.1% NaN3 | |
ITGB1, GPIIA, OTTHUMP00000063731, OTTHUMP00000046253, OTTHUMP00000063733, VLAB, OTTHUMP00000063732, OTTHUMP00000046253, MDF2, FNRB, CD29, MSK12, VLA-BETA | |
Protein A purfied | |
Primary | |
2-8°C for one year | |
IgG2 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction