Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin Β1 Mouse anti-Human, Clone: N29, MilliporeSigma™

Catalog No. MAB2252ZIMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2252ZIMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2252ZIMI Supplier MilliporeSigma Supplier No. MAB2252ZI
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Integrin β1 Antibody, clone N29 (Azide Free) is validated for use in Flow Cytometry and Immunohistochemistry and Immunoprecipitation and Affects Function and Cell Culture and Western Blotting for the detection of Integrin β1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin β1
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone N29
Conjugate Unconjugated
Formulation Purified cultured supernatant in PBS without preservatives.
Gene Accession No. NM_002292.3
Gene Symbols LAMB2, S-laminin, LAMS
Host Species Mouse
Immunogen Jurkat T-leukemic cell line
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.