Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin Β1 Mouse anti-Human, Clone: P5D2, MilliporeSigma™

Catalog No. MAB1959MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1959MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1959MI Supplier MilliporeSigma Supplier No. MAB1959
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Integrin β1 Antibody, clone P5D2 detects level of Integrin β1 and has been published and validated for use in ELISA, flow cytometry, immunocytochemistry, immunohistochemistry and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin β1
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone P5D2
Conjugate Unconjugated
Formulation Purified immunoglobulin in 0.02M PB, 0.25M NaCl, pH 7.6, 0.1% sodium azide.
Gene Accession No. NM_002211.2; NM_033666.1; NM_033667.1; NM_033668.1; NM_033669.1; NM_133376.1
Gene Symbols ITGB1, GPIIA, OTTHUMP00000063731, OTTHUMP00000046253, OTTHUMP00000063733, VLAB, OTTHUMP00000063732, MDF2, FNRB, CD29, MSK12
Host Species Mouse
Immunogen Human keratinocytes derived from skin.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.