Learn More
MilliporeSigma™ anti-Integrin β1, Cytosolic (CD29); Polyclonal; ™ (Chemicon™)
Supplier:  MilliporeSigma™ AB1952/DEL
Catalog No. 50-173-332
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
Description
Description
Immunoprecipitation, Western Blotting
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Integrin β1, Cytosolic (CD29)
Unconjugated
Synthetic peptide corresponding to the c-terminal cytoplasmic domain of human β1 integrin.
RUO
Primary
Serum
Polyclonal
Rabbit
100 μL
Cell Structure
Human, Mouse, Pig, Rat
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.