Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin Β1 Mouse anti-Human, FITC, Clone: HUTS-4, MilliporeSigma™

Catalog No. FCMAB389FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB389FMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB389FMI Supplier MilliporeSigma Supplier No. FCMAB389F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Milli-Mark™ Anti-Integrin β1-FITC Antibody, activated, clone HUTS-4 is an antibody against Integrin β1 for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin β1
Applications Flow Cytometry
Classification Monoclonal
Clone HUTS-4
Conjugate FITC
Formulation Purified mouse monoclonal IgG2b conjugated to FITC in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Accession No. NP_002202
Gene Symbols LAMB2, S-laminin, LAMS
Host Species Mouse
Immunogen Generated against human beta1 integrins purified from Triton X-100 lysates of human lung, liver, and skeletal muscle tissues.
Purification Method Protein A Purified
Quantity 100 Tests
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from Light.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.