Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin Β1 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB1929
Description
Anti-Integrin β1 Antibody, phospho-specific [Thr788/Thr789] is an antibody against Integrin β1 for use in western blotting.
Specifications
| Integrin β1, phospho-specific [Thr788/Thr789] | |
| Polyclonal | |
| Antibody is cross-adsorbed to nonphospho-peptide corresponding to the site of phosphorylation then affinity purified using phosphorylated integrin beta1 receptor (Thr788/789) peptide. Antibody is supplied in Dulbecco's phosphate-buffered saline (without M | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Human |
| Western Blot | |
| Unconjugated | |
| ITGB1; GPIIA; OTTHUMP00000063731; OTTHUMP00000046253; OTTHUMP00000063733; VLAB; OTTHUMP00000063732; MDF2; FNRB; CD29; MSK12 | |
| Phosphorylated synthetic peptide corresponding to amino acid residues surrounding and including threonine 788/789 of the human beta1 integrin. The immunogen sequence is 100% identical in mouse, rat and chicken integrin beta1. | |
| 100 μL | |
| NM_002211.2; NM_033666.1; NM_033667.1; NM_033668.1; NM_033669.1; NM_133376.1 | |
| −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction