Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin β1 Rabbit anti-Human, Mouse, Pig, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB1952MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1952MI 100 μL
AB1952PMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AB1952MI Supplier MilliporeSigma Supplier No. AB1952
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Integrin beta1 Antibody, Cytosolic detects level of Integrin beta1 and has been published and validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin beta1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum containing no preservatives
Gene Symbols ITGB1; GPIIA; VLAB; MDF2; FNRB; CD29; MSK12
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the c-terminal cytoplasmic domain of human β-1 integrin.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002211.2
Target Species Human, Mouse, Pig, Rat
Content And Storage −20°C for up to 12 months
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.