Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin Β1D Mouse anti-Human, Mouse, Canine, Clone: 2B1, MilliporeSigma™

Catalog No. MAB1900MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1900MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1900MI Supplier MilliporeSigma Supplier No. MAB1900
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Integrin β1D Antibody, C-terminus cytoplasmic domain, clone 2B1 is validated for use in western blot, immunocytochemistry, immunohistochemistry for the detection of Integrin β1D.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin β1D
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2B1
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in PBS containing 0.09% sodium azide.
Gene Accession No. NM_002211.2; NM_033666.1; NM_033667.1; NM_033668.1; NM_033669.1; NM_133376.1
Gene Symbols ITGB1, GPIIA, OTTHUMP00000063731, OTTHUMP00000046253, OTTHUMP00000063733, VLAB, OTTHUMP00000063732, MDF2, FNRB, CD29, MSK12
Host Species Mouse
Immunogen Synthetic peptide including an appending N-terminal cysteine: CQENPIYKSPINNFKNPNYGRKAGL coupled to KLH.
Quantity 100 μg
Primary or Secondary Primary
Target Species Canine, Human, Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.