Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Integrin α2Β1 Mouse anti-Human, Pig, Clone: BHA2.1

Catalog No. MAB1998ZMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1998ZMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1998ZMI Supplier MilliporeSigma™ Supplier No. MAB1998Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Integrin α2β1 Antibody, clone BHA2.1, azide free is validated for use in flow cytometry, immunohistochemistry, immunoprecipitation, FUNC for the detection of Integrin α2β1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin α2β1
Applications Flow Cytometry, Functional Assay, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone BHA2.1
Conjugate Unconjugated
Formulation Purified immunoglobulin from Protein A chromatography. Liquid in 0.02M PB pH 7.6, 0.25M NaCl containing no preservative.
Gene Accession No. NM_002203.3
Gene Symbols ITGA2, CD49B, VLAA2, VLA-2, CD49b, BR, GPIa
Host Species Mouse
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Pig
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.