Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin α2Β1 Rat anti-Mouse, Clone: BMA2.1, MilliporeSigma™

Catalog No. MAB2141ZMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2141ZMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2141ZMI Supplier MilliporeSigma Supplier No. MAB2141Z

Rat Monoclonal Antibody

Detect Integrin α2β1 using this Anti-Integrin α2β1 Antibody, clone BMA2.1, azide free validated for use in FC and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin α2β1
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone BMA2.1
Conjugate Unconjugated
Formulation Liquid containing no preservatives.
Gene Accession No. NM_002203.3
Gene Symbols ITGA2, CD49B, VLAA2, VLA-2, CD49b, BR, GPIa
Host Species Rat
Immunogen Mouse integrin alpha2beta1 heterodimer
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.