Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin Β2 [CD18] Mouse anti-Human, Clone: MEM 48, MilliporeSigma™

Catalog No. CBL158MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL158MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL158MI Supplier MilliporeSigma Supplier No. CBL158
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Integrin β2 [CD18] Antibody detects level of Integrin β2 [CD18] and has been published and validated for use in flow cytometry, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin β2 [CD18]
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone MEM 48
Conjugate Unconjugated
Formulation The monoclonal is presented in phosphate buffered saline containing 15mM sodium azide, pH 7.4. We recommend that each laboratory determine an optimum working titre for use in its particular application.
Gene Accession No. NM_000211.2
Gene Symbols ITGB2, MFI7, LAD, CD18, MF17, LCAMB, LFA-1, MAC-1
Host Species Mouse
Immunogen Leucocytes from an LGL - type leukaemia
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.