Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin Β2 Mouse anti-Human, Clone: P4H9, MilliporeSigma™

Catalog No. MAB1962MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1962MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1962MI Supplier MilliporeSigma Supplier No. MAB1962
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Integrin β2 using this Anti-Integrin β2 Antibody, clone P4H9 validated for use in immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin β2
Applications Immunohistochemistry
Classification Monoclonal
Clone P4H9
Conjugate Unconjugated
Formulation Non-sterile buffer of 0.02M phosphate, 0.25M Sodium Chloride, pH 7.6,

0.1% sodium azide.
Gene Accession No. NM_000211.2
Gene Symbols ITGB2, LFA-1, MAC-1, MF17, LCAMB, MFI7, LAD, CD18
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C.
Form Purified
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.