Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin α3Β1 Mouse anti-Human, Clone: M-KID2, MilliporeSigma™

Catalog No. MAB1992MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1992MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1992MI Supplier MilliporeSigma Supplier No. MAB1992
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Integrin α3β1 Antibody, clone M-KID2 detects level of Integrin α3β1 and has been published and validated for use in ELISA, immunoprecipitation, RIA, immunocytochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin α3β1
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone M-KID2
Conjugate Unconjugated
Formulation Immunoglobulin purified by Protein A. Purified immunoglobulin in PBS and 0.05% sodium azide as preservative.
Gene Accession No. NM_002204.1; NM_005501.1
Gene Symbols ITGA3, GAP-B3, CD49c, FLJ34704, MSK18, VLA3a, VL3A, VCA-2, FLJ34631, FRP-2, GAPB3, CD49C
Host Species Mouse
Immunogen Human renal carcinoma cell line (KJ29).
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.