Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin Β3 (CD61) Mouse anti-Human, Clone: SAP, MilliporeSigma™

Catalog No. 05769MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-769-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-769-MI Supplier MilliporeSigma Supplier No. 05769
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Integrin β3 (CD61) Antibody, clone SAP detects level of Integrin β3 (CD61) and has been published and validated for use in western blot, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin β3 (CD61)
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone SAP
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, and 0.05% sodium azide.
Gene Accession No. NM_000212
Gene Symbols ITGB3, GP3A, GPIIIa, CD61
Host Species Mouse
Immunogen Human platelet membrane fractions
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.