Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin Β3 Mouse anti-Human, Clone: 25E11, MilliporeSigma™

Catalog No. MAB1957ZMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1957ZMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1957ZMI Supplier MilliporeSigma Supplier No. MAB1957Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Integrin β3 using this Anti-Integrin β3 Antibody, clone 25E11, azide free validated for use in flow cytometry, immunoprecipitation and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin β3
Applications Flow Cytometry, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 25E11
Conjugate Unconjugated
Formulation In buffer of 0.01M phosphate, 0.15M sodium chloride, pH 7.1, containing no preservatives.
Gene Accession No. NM_000212.2
Gene Symbols ITGB3, GP3A, CD61, GPIIIa
Host Species Mouse
Immunogen Normal blood mononuclear cells activated by mixed lymphocyte reaction and cultured for 10 days in medium containing IL-2.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.