Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin α3 Mouse anti-Human, Clone: ASC-1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB2056Z
Description
Anti-Integrin α3 Antibody, clone ASC-1 (Azide Free) inhibits cell attachment and detects level of Integrin α3 and has been published and validated for use in flow cytometry, immunoprecipitation, immunohistochemistry, Inhibition Assays.
Specifications
Integrin α3 | |
Monoclonal | |
Unconjugated | |
NM_002204.1; NM_005501.1 | |
Mouse | |
100 μg | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Inhibition Assays | |
ASC-1 | |
Purified cultured supernatant in 0.1MTris-Glycine (pH 7.4), 150mM NaCl, without preservatives. | |
ITGA3, GAP-B3, CD49c, FLJ34704, MSK18, VLA3a, VL3A, VCA-2, FLJ34631, FRP-2, GAPB3, CD49C | |
Human squamous cell carcinoma cells (SCC9) | |
Primary | |
−20°C in undiluted aliquots. Avoid repeated freeze/thaw cycles. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction