Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin Β3 Mouse anti-Human, Clone: B3A, MilliporeSigma™

Catalog No. MAB2023ZMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2023ZMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2023ZMI Supplier MilliporeSigma Supplier No. MAB2023Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Integrin β3 Antibody, clone B3A, azide free detects level of Integrin β3 and has been published and validated for use in flow cytometry, FUNC and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin β3
Applications Flow Cytometry, Functional Assay, Western Blot
Classification Monoclonal
Clone B3A
Conjugate Unconjugated
Formulation Protein A Purified mouse immunoglobulin in 20mM sodium phosphate, 250mM NaCl, pH. 7.6, with no preservatives.
Gene Accession No. NM_000212.2
Gene Symbols ITGB3, GP3A, CD61, GPIIIa
Host Species Mouse
Immunogen Human integrin alphaVbeta3 heterodimer isolated from placenta.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.