Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin α3 Rabbit anti-Bovine, Canine, Chicken, Goat, Hamster, Horse, Human, Mouse, Pig, Rat, Sheep, Polyclonal, MilliporeSigma™

Catalog No. AB1920MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1920MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1920MI Supplier MilliporeSigma Supplier No. AB1920
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Integrin α3 using this Anti-Integrin α3 Antibody validated for use in ELISA, immunofluoresence, immunohistochemistry, immunoprecipitation, RIA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin α3
Applications ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal IgG antiserum containing 0.05% sodium azide.
Gene Symbols ITGA3; GAP-B3; CD49c; FLJ34704; MSK18; VLA3a; VL3A; VCA-2; FLJ34631; FRP-2; GAPB3; CD49C
Host Species Rabbit
Immunogen A synthetic peptide (CRIQPSETERLTDDY) derived from the COOH terminal sequence located in the cytoplasmic domain of the human alpha3 integrin subunit.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002204.1
Target Species Bovine, Canine, Chicken, Equine, Goat, Hamster, Human, Mouse, Pig, Rat, Sheep
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.