Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin Β4 Mouse anti-Human, Clone: ASC-8, MilliporeSigma™

Catalog No. MAB2059MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2059MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2059MI Supplier MilliporeSigma Supplier No. MAB2059
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Integrin β4 Antibody, clone ASC-8 is an antibody against Integrin β4 for use in flow cytometry, immunoprecipitation, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin β4
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone ASC-8
Conjugate Unconjugated
Formulation Liquid in 0.02M PB pH 7.6, 0.25M NaCl containing 0.1% sodium azide.
Gene Accession No. NM_000213.3; NM_001005619.1; NM_001005731.1
Gene Symbols ITGB4, CD104, GP150
Host Species Mouse
Immunogen Human squamous cell carcinoma cells (SCC9)
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.