Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin α4 Mouse anti-Bovine, Human, Rat, Clone: HP2/1, MilliporeSigma™

Catalog No. MAB1383MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1383MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1383MI Supplier MilliporeSigma Supplier No. MAB1383
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Integrin α4 using this Anti-Integrin α4 Antibody, clone HP2/1 validated for use in flow cytometry, immunoprecipitation, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin α4
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone HP2/1
Conjugate Unconjugated
Formulation Purified immunoglobulin - Ig fraction. Liquid in Phosphate Buffered Saline, pH 7.4 with 0.1% Sodium Azide.
Gene Accession No. NM_000885.4
Gene Symbols ITGA4, MGC90518, VLA-4, IA4, CD49D, CD49d
Host Species Mouse
Immunogen JM leukemia line
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Human, Rat
Content And Storage −20°C in undiluted aliquots. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.