Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin α4 Mouse anti-Human, Primate, Clone: P1H4, MilliporeSigma™

Catalog No. MAB16983MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB16983MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB16983MI Supplier MilliporeSigma Supplier No. MAB16983
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Integrin α4 Antibody, clone P1H4 detects level of Integrin α4 and has been published and validated for use in ELISA, flow cytometry, immunoprecipitation, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin α4
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone P1H4
Conjugate Unconjugated
Formulation Purified immunoglobulin from Protein A Sepharose™ chromatography. Liquid in 0.02M PB, pH 7.6, 0.25M NaCl containing 0.01% sodium azide.
Gene Accession No. NP_000876.3
Gene Symbols CD49D, ITGA4
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Primate
Content And Storage 2-8°C.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.