Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin α4 Mouse anti-Human, Clone: P4C2, MilliporeSigma™

Catalog No. MAB1955MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1955MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1955MI Supplier MilliporeSigma Supplier No. MAB1955
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Integrin α4 Antibody, clone P4C2 is validated for use in immunoprecipitation, immunohistochemistry, FUNC for the detection of Integrin α4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin α4
Applications Functional Assay, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone P4C2
Conjugate Unconjugated
Formulation Liquid ascites containing sodium azide as a preservative.
Gene Accession No. NM_000885.4
Gene Symbols ITGA4, MGC90518, VLA-4, IA4, CD49D, CD49d
Host Species Mouse
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.