Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin Β4 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB1922
Description
Anti-Integrin β4 Antibody is an antibody against Integrin β4 for use in immunoprecipitation, immunohistochemistry.
Specifications
Integrin β4 | |
Polyclonal | |
Whole rabbit serum. Liquid, with 0.02% sodium azide. | |
Rabbit | |
100 μL | |
NM_000213.3; NM_001005619.1; NM_001005731.1 | |
-20°C in undiluted aliquotes for up to 6 months, Avoid Freeze/Thaw Cycles |
Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
ITGB4; CD104; GP150 | |
Synthetic peptide derived from the COOH terminal sequence (cytoplasmic domain) of the human β4 integrin subunit (Tamura et al. 1990). The sequence used is NH2-(K)GTLSTHMDQQFFQT-amide. The lysine (K) is not in the β4 sequence and was added to con | |
Primary | |
Human, Mouse, Rat | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction