Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin α5Β1 Rat anti-Mouse, Clone: BMB5, MilliporeSigma™

Catalog No. MAB2514MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2514MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2514MI Supplier MilliporeSigma Supplier No. MAB2514
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This Anti-Integrin α5β1 Antibody, clone BMB5 is validated for use in flow cytometry, immunoprecipitation, western blot, immunohistochemistry, FUNC for the detection of Integrin α5β1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin α5β1
Applications Flow Cytometry, Functional Assay, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone BMB5
Conjugate Unconjugated
Formulation Liquid, containing no preservatives.
Gene Accession No. NM_002205.2
Gene Symbols ITGA5, VLA-5, CD49e, VLA5A, FNRA
Host Species Rat
Immunogen The antibody was prepared by fusion of spleen cells from a Fisher rat immunized with mouse embryonal carcinoma P19 cells with myeloma P3X63Ag9.653 cells.
Quantity 100 μL
Primary or Secondary Primary
Target Species Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months.
Form Ascites
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.