Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin α5 Rabbit anti-Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB1921
Description
Detect Integrin α5 using this Anti-Integrin α5 Antibody validated for use in ELISA, western blot, immunohistochemistry.
Specifications
Integrin α5 | |
Polyclonal | |
Whole rabbit serum. Liquid, with 0.02% sodium azide. | |
Rabbit | |
100 μL | |
NM_002205.2 | |
−20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles. |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
ITGA5; VLA-5; CD49e; VLA5A; FNRA | |
Recombinant fragment of mouse alpha5 integrin (Holers et al. 1989) corresponding to amino acid residues 655-880 of the human protein (Argraves 1987). This region of the molecule is extracellular and is included in the heavy chain (120 kDa). | |
Primary | |
Mouse | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction