Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin Β5 Rabbit anti-Human, Hamster, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB1926MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1926MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1926MI Supplier MilliporeSigma Supplier No. AB1926
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Integrin β5 using this Anti-Integrin β5 Antibody, C-terminus/cytosolic validated for use in ELISA, immunofluoresence, immunohistochemistry (paraffin), immunoprecipitation, RIA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin β5
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation UnPurified whole rabbit Serum containing 0.05% sodium azide as a preservative.
Gene Symbols ITGB5; FLJ26658
Host Species Rabbit
Immunogen Synthetic peptide derived from the COOH terminal sequence (cytoplasmic domain) of the human integrin beta 5 subunit (KTFNKFNKSYNGTVD). Note : a lysine (K) residue was added to the N-terminal of the peptide to facilitate coupling to the carrier protein.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002213.3
Target Species Hamster, Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.