Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Integrin Β6 Mouse anti-Human, Monkey, Rat, Rabbit, Clone: CSbeta6

Catalog No. MAB2076ZMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2076ZMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2076ZMI Supplier MilliporeSigma™ Supplier No. MAB2076Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Integrin β6 Antibody, clone CSβ6, azide free detects level of Integrin β6 and has been published and validated for use in flow cytometry, immunoprecipitation, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin β6
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone CSbeta6
Conjugate Unconjugated
Formulation Sterile-filtered liquid at 1mg/mL in PBS, containing no preservatives.
Gene Accession No. NM_000888.3
Gene Symbols ITGB6
Host Species Mouse
Immunogen beta 6 knockout mice have been immunized with recombinant secreted human alpaVbeta6.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Monkey, Rabbit, Rat
Content And Storage 2-8°C.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.