Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin α6 Rat anti-Human, Mouse, Clone: NKI-GoH3, MilliporeSigma™

Catalog No. MAB1378MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1378MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1378MI Supplier MilliporeSigma Supplier No. MAB1378
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Detect Integrin α6 using this Anti-Integrin α6 Antibody, clone NKI-GoH3 validated for use in flow cytometry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin α6
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone NKI-GoH3
Conjugate Unconjugated
Formulation Protein A Purified mouse immunoglobulin in 20mM sodium phosphate, 250mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative.
Gene Accession No. NM_000210.2; NM_001079818.1
Gene Symbols ITGA6, VLA-6, ITGA6B, CD49f
Host Species Rat
Immunogen Mammary tumor cells from BALB/c mice.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.