Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin Β6 Mouse anti-Human, Clone: R6G9, MilliporeSigma™

Catalog No. MAB2075ZMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2075ZMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2075ZMI Supplier MilliporeSigma Supplier No. MAB2075Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Integrin β6 Antibody, clone R6G9, azide free is an antibody against Integrin β6 for use in FC and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin β6
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone R6G9
Conjugate Unconjugated
Formulation Sterile-filtered liquid at 1mg/mL in PBS, containing no preservatives.
Gene Accession No. NM_000888.3
Gene Symbols ITGB6
Host Species Mouse
Immunogen Murine NIH 3T3 cells stably transfected with the human (6 subunit
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.