Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha-11 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB6031
Description
Anti-Integrin alpha-11 Antibody is an antibody against Integrin alpha-11 for use in western blotting.
Specifications
Integrin alpha-11 | |
Polyclonal | |
Purified rabbit polyclonal containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat |
Western Blot | |
Unconjugated | |
ITGA11 | |
Linear peptide corresponding to human Integrin alpha-11. | |
100 μg | |
NP_001004439 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction