Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Integrin alpha L/CD11a Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3595
This item is not returnable.
View return policy
Description
Integrin alpha L/CD11a Monoclonal specifically detects Integrin alpha L/CD11a in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
Integrin alpha L/CD11a | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
CAA68747 | |
ITGAL | |
Chinese hamster ovary cell line CHO-derived recombinant human Integrin alpha L Tyr26-Met1089 (Tyr660Ile) Accession # CAA68747 | |
100 μg | |
Primary | |
Detects human Integrin alpha L in Western blots. | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
345913 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD11 antigen-like family member A, CD11a, CD11a antigen, CD11Aantigen CD11A (p180), lymphocyte function-associated antigen 1, alphapolypeptide, integrin alpha-L, integrin gene promoter, integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen1; alpha polypeptide), ITGAL, Leukocyte adhesion glycoprotein LFA-1 alpha chain, Leukocyte function-associated molecule 1 alpha chain, LFA-1, LFA-1 alpha, LFA1A, LFA-1A, Ly-15, Ly-21, lymphocyte function-associated antigen 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3683 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction