Learn More
Human Integrin alpha M/CD11b Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Supplier:  R&D Systems FAB16991T025
 View more versions of this product
Catalog No. FAB16991T02
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
CD11b Monoclonal specifically detects CD11b in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD11b
Monoclonal
Alexa Fluor 594
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, Integrin alpha M, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), ITGAM, Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
3684
Human
Supernatant
Flow Cytometry
238446
Flow Cytometry 5 uL/10^6 cells
NP_001139280
ITGAM
Mouse myeloma cell line NS0-derived recombinant human Integrin alpha M/CD11b Phe17-Asn1105 Accession # NP_001139280
25 Tests
Primary
Detects human Integrin alpha M/CD11b in direct ELISAs.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only