Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alphaV Rabbit anti-Chicken, Horse, Goat, Human, Mouse, Hamster, Rat, Pig, Sheep, Polyclonal, MilliporeSigma™

Catalog No. AB1930MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1930MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1930MI Supplier MilliporeSigma Supplier No. AB1930
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Integrin alphaV Antibody, C-terminus, Intracellular detects level of Integrin alphaV and has been published and validated for use in ELISA, immunohistochemistry, immunoprecipitation, RIA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin alphaV
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Neat rabbit antiserum in liquid with 0.05% sodium azide.
Gene Symbols ITGAV; DKFZp686A08142; MSK8; CD51; VNRA
Host Species Rabbit
Immunogen Synthetic peptide derived from the COOH terminal sequence (cytoplasmic domain) of the human alphaV integrin subunit (SwissProt Accession P06756, amino acids 1022-1034).
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002210.2
Target Species Chicken, Equine, Goat, Hamster, Human, Mouse, Pig, Rat, Sheep
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.