Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Integrin beta1 Alexa Fluor 488, Clone: HUTS-4,

Catalog No. MAB2079AF48 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 488
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MAB2079AF48 100 μL Alexa Fluor 488
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2079AF48 Supplier MilliporeSigma™ Supplier No. MAB2079AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Integrin β1 Clone: HUTS-4 in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry

Integrin beta-1 (UniProt P05556; also known as CD29, Glycoprotein Iia, GPIIA, Integrin VLA-4 subunit beta, Very late activation protein, beta polypeptide) is encoded by the ITGB1 (also known as FNRB, MDF2, MSK12, VLA-BETA, VLAB) gene (Gene ID 3688) in human. Integrin beta-1 is a 130kDa transmembrane glycoprotein that interact with various integrin alpha subunits (including alpha 1, alpha 2, alpha 3, alpha 4, alpha 5, and alpha 6) to form the functional receptor complexes that bind to specific extracellular matrix proteins. Integrin receptors regulate a variety of important biological functions, including embryonic development, wound repair, hemostasis, and prevention of programmed cell death. They are also implicated in abnormal pathological states such as tumor directed angiogenesis, tumor growth and metastasis. These heterodimeric receptors bridge the cytoplasmic actin cytoskeleton with proteins present in the extracellular matrix and/or on adjacent cells. Interactions between integrins and the extracellular matrix lead to the activation of signal transduction pathways and regulation of gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin β1
Applications Immunocytochemistry
Classification Monoclonal
Clone HUTS-4
Conjugate Alexa Fluor 488
Formulation Purified mouse antibody conjugate in PBS with 15mg/ml BSA and 0.1 % sodium azide.
Gene Accession No. P05556
Gene Symbols ITGB1, FNRB, MDF2, MSK12
Host Species Mouse
Immunogen Generated against human β1 integrins purified from Triton X-100 lysates of human lung, liver, and skeletal muscle tissues.
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NM_002292.3
Test Specificity Clone HUTS-4 is specific for active conformation of human beta1 integrins. The beta1 or VLA integrin subgroup contains at least 10 members, each with a distinct alpha subunit nonconvalently associated with the common beta1 subunit. Functional activation of integrins can be experimentally induced by addition of Mg2+ or Mn2+, or by using one of several stimulatory antibodies that are available. Intracellular signals generated as a result of cell activation also modulate the confirmation and affinity of integrins in cellular systems. Antibodies which have the ability to discriminate between different states of integrin activation are exceptionally useful for studies on the regulation of activation. Recognizes epitopes in the 355-425 region of the common beta1 subunit of VLA integrins, whose expression parallels the ligand binding activity of these adhesion receptors induced by various extracellular and intracellular stimuli.
Target Species Human, Mouse, Rat
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.