Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin Beta1 Mouse anti-Human, Clone: 8E3, MilliporeSigma™

Catalog No. MABT199MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT199MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT199MI Supplier MilliporeSigma Supplier No. MABT199
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Integrin beta1 Antibody is validated for use in western blot, immunoprecipitation and activity assays for the detection of Integrin beta1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin Beta1
Applications Functional Assay, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 8E3
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing PBS without preservatives.
Gene Accession No. NP_002202
Gene Symbols ITGB, FNRB, MDF2, MSK12
Host Species Mouse
Immunogen Purified Integrin Beta1 from HT-1080 cells.
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.