Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta1 Hamster anti-Human, Mouse, Clone: HM beta 1.1, MilliporeSigma™

Catalog No. CBL1348MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL1348MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL1348MI Supplier MilliporeSigma Supplier No. CBL1348

Hamster Monoclonal Antibody

Detect Integrin beta1 using this Anti-Integrin beta1 Antibody, clone HM beta 1.1 validated for use in immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin Beta1
Applications Immunocytochemistry
Classification Monoclonal
Clone HM beta 1.1
Conjugate Unconjugated
Formulation Purified in PBS with 0.05% NaN3.
Gene Accession No. NP_002202.2
Gene Symbols CD29, FNRB, GPIIA , MDF2 , MSK12 , OTTHUMP00000019420 , OTTHUMP00000046253, OTTHUMP00000063731, OTTHUMP00000063732, OTTHUMP00000063733, VLA-BETA, VLAB
Host Species Hamster
Immunogen Purified mouse VLA-4 antigen
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.