Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Integrin beta6 Clone: 442.5C4

Catalog No. 407317100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
40-731-7100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-731-7100UL Supplier MilliporeSigma™ Supplier No. 407317100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Integrin β6 Clone: 442.5C4 in Human samples, and it is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immuno Blotting

The integrins are a family of more than 23 heterodimeric transmembrane proteins that mediate cell-cell adhesions as well as cell-substratum adhesions and signal transduction processes. Integrins are heterodimers consisting of noncovalently associated α and β subunits. Integrin β6 has been shown to play a role in cell spreading, migration, and growth during reorganization of epithelia in development, tissue repair, and neoplasia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin β6
Applications ELISA, Flow Cytometry, Immunoblot, Immunoblot, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone 442.5C4
Conjugate Unconjugated
Formulation Undiluted ascites.
Host Species Mouse
Immunogen recombinant, human αvβ6
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the β6 integrin chain.
Target Species Human
Content And Storage −20°C
Form Ascites
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.